3-Fluoropicolinaldehyde hydrochloride ≥97% research grade

Supplier: AOBChem USA

Warning

38293-25G 38293-5G
76915-964EA 275.84 USD
76915-964 76915-966
3-Fluoropicolinaldehyde hydrochloride ≥97% research grade
3-Fluoropicolinaldehyde hydrochloride
Formula: C₆H₅ClFNO
MW: 161.561 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32632325
CAS Number: 2404733-71-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR