3,5-Difluoro-4-propoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

38261-1G 38261-25G 38261-500MG 38261-5G
76915-950EA 231.28 USD
76915-950 76915-952 76915-954 76915-956
3,5-Difluoro-4-propoxybenzoic acid ≥97%
3,5-Difluoro-4-propoxybenzoic acid
Formula: C₁₀H₁₀F₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD18285395
CAS Number: 1342624-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR