4,6-Dibromo-3-ethoxy-2-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

38164-1G 38164-25G 38164-500MG 38164-5G
76915-926EA 231.28 USD
76915-926 76915-928 76915-930 76915-932
4,6-Dibromo-3-ethoxy-2-fluorobenzoic acid ≥97%
4,6-Dibromo-3-ethoxy-2-fluorobenzoic acid
Formula: C₉H₇Br₂FO₃
MW: 341.956 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32632312
CAS Number: 2404734-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR