3-Bromo-N-cyclohexyl-5-nitrobenzamide ≥97%

Supplier: AOBChem USA

Warning

38070-25G 38070-5G
76915-914EA 445.58 USD
76915-914 76915-916
3-Bromo-N-cyclohexyl-5-nitrobenzamide ≥97%
3-Bromo-N-cyclohexyl-5-nitrobenzamide

Precaución: for Research Use Only

Formula: C₁₃H₁₅BrN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD09475857
CAS Number: 941294-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR