3-Bromo-N-cyclopropyl-5-nitrobenzamide ≥97%

Supplier: AOBChem USA

Warning

38068-25G 38068-5G
76915-904EA 559.1 USD
76915-904 76915-906
3-Bromo-N-cyclopropyl-5-nitrobenzamide ≥97%
3-Bromo-N-cyclopropyl-5-nitrobenzamide

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD09475856
CAS Number: 941294-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR