3,5-Dimethyl-4-propoxybenzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

38002-1G 38002-25G 38002-500MG 38002-5G
76915-866EA 236.59 USD
76915-866 76915-868 76915-870 76915-872
3,5-Dimethyl-4-propoxybenzaldehyde ≥97% research grade
3,5-Dimethyl-4-propoxybenzaldehyde
Formula: C₁₂H₁₆O₂
Storage Temperature: Refrigerator
MDL Number: MFCD06247276
CAS Number: 210057-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR