4-(Benzyloxy)-5-bromo-2,3-difluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

37886-1G 37886-500MG 37886-5G
76915-836EA 498.63 USD
76915-836 76915-838 76915-840
4-(Benzyloxy)-5-bromo-2,3-difluorobenzoic acid ≥97%
4-(Benzyloxy)-5-bromo-2,3-difluorobenzoic acid
Formula: C₁₄H₉BrF₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD32632270
CAS Number: 2404734-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR