4-(Thiophen-2-yl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

37767-25G 37767-5G
76915-824EA 573.95 USD
76915-824 76915-826
4-(Thiophen-2-yl)benzaldehyde ≥95%
4-(Thiophen-2-yl)benzaldehyde
Formula: C₁₁H₈OS
Storage Temperature: Ambient
MDL Number: MFCD02682003
CAS Number: 107834-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR