4-Bromo-2-chloro-5-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

37603-1G 37603-25G 37603-500MG 37603-5G
76915-778EA 290.69 USD
76915-778 76915-780 76915-782 76915-784
4-Bromo-2-chloro-5-methoxyphenol ≥97%
4-Bromo-2-chloro-5-methoxyphenol
Formula: C₇H₆BrClO₂
MW: 237.478 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29044460
CAS Number: 1881328-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR