4,4,5,5-Tetramethyl-2-(4-(2,5-dimethylpyrrol-1-yl)phenyl)-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

37225-1G 37225-500MG 37225-5G
76915-752EA 528.33 USD
76915-752 76915-754 76915-756
4,4,5,5-Tetramethyl-2-(4-(2,5-dimethylpyrrol-1-yl)phenyl)-1,3,2-dioxaborolane ≥97%
4,4,5,5-Tetramethyl-2-(4-(2,5-dimethylpyrrol-1-yl)phenyl)-1,3,2-dioxaborolane
Formula: C₁₈H₂₄BNO₂
MW: 297.2 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31916438
CAS Number: 1421275-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR