3-Chloro-4-ethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

37194-25G 37194-5G
76915-740EA 466.8 USD
76915-740 76915-742
3-Chloro-4-ethoxybenzoic acid ≥97%
3-Chloro-4-ethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉ClO₃
MW: 200.619 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02063066
CAS Number: 213598-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR