3,6-Difluoro-2-iodophenol ≥97%

Supplier: AOBChem USA

Warning

37186-1G 37186-500MG 37186-5G
76915-728EA 457.25 USD
76915-728 76915-730 76915-732
3,6-Difluoro-2-iodophenol ≥97%
3,6-Difluoro-2-iodophenol
Formula: C₆H₃F₂IO
MW: 255.989 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632154
CAS Number: 2092374-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR