3-Fluoro-2-methoxy-6-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

37042-1G 37042-500MG 37042-5G
76915-704EA 492.26 USD
76915-704 76915-706 76915-708
3-Fluoro-2-methoxy-6-methylbenzaldehyde ≥97%
3-Fluoro-2-methoxy-6-methylbenzaldehyde
Formula: C₉H₉FO₂
Storage Temperature: Refrigerator
MDL Number: MFCD22415109
CAS Number: 1781804-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR