3,4-Dibromo-2-fluorobenzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

36816-1G 36816-25G 36816-500MG 36816-5G
76915-678EA 324.64 USD
76915-678 76915-680 76915-682 76915-684
3,4-Dibromo-2-fluorobenzaldehyde ≥97% research grade
3,4-Dibromo-2-fluorobenzaldehyde
Formula: C₇H₃Br₂FO
Storage Temperature: Refrigerator
MDL Number: MFCD28784591
CAS Number: 1804931-57-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR