3',5'-Dimethyl-[1,1'-biphenyl]-4-carbonitrile ≥97%

Supplier: AOBChem USA

Warning

36771-1G 36771-25G 36771-5G
76915-664EA 96.55 USD
76915-664 76915-666 76915-668
3',5'-Dimethyl-[1,1'-biphenyl]-4-carbonitrile ≥97%
3',5'-Dimethyl-[1,1'-biphenyl]-4-carbonitrile
Formula: C₁₅H₁₃N
MW: 207.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21609633
CAS Number: 935552-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR