4-Bromo-2-(methoxymethyl)fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

36725-1G 36725-25G 36725-500MG 36725-5G
76915-648EA 203.7 USD
76915-648 76915-650 76915-652 76915-654
4-Bromo-2-(methoxymethyl)fluorobenzene ≥97%
4-Bromo-2-(methoxymethyl)fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₈BrFO
Storage Temperature: Refrigerator
MDL Number: MFCD04116325
CAS Number: 337535-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR