4-Bromo-2-(trifluoromethyl)benzoyl chloride ≥95%

Supplier: AOBChem USA

Warning

36719-25G 36719-5G
76915-638EA 310.85 USD
76915-638 76915-640
4-Bromo-2-(trifluoromethyl)benzoyl chloride ≥95%
4-Bromo-2-(trifluoromethyl)benzoyl chloride
Formula: C₈H₃BrClF₃O
Storage Temperature: Refrigerator
MDL Number: MFCD13185394
CAS Number: 104356-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR