3-Fluoro-2-methoxy-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

36675-1G 36675-500MG 36675-5G
76915-630EA 528.33 USD
76915-630 76915-632 76915-634
3-Fluoro-2-methoxy-5-methylbenzoic acid ≥97%
3-Fluoro-2-methoxy-5-methylbenzoic acid
Formula: C₉H₉FO₃
Storage Temperature: Refrigerator
MDL Number: MFCD23712102
CAS Number: 1427427-96-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR