3-Bromo-5-(difluoromethyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

36454-1G 36454-25G 36454-500MG 36454-5G
76915-590EA 122.01 USD
76915-590 76915-592 76915-594 76915-596
3-Bromo-5-(difluoromethyl)pyridine ≥97%
3-Bromo-5-(difluoromethyl)pyridine
Formula: C₆H₄BrF₂N
MW: 208.006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16140196
CAS Number: 114468-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR