3,5-Difluoro-4-methylphenol ≥97%

Supplier: AOBChem USA

Warning

35836-1G 35836-500MG 35836-5G
76915-524EA 680.04 USD
76915-524 76915-526 76915-528
3,5-Difluoro-4-methylphenol ≥97%
3,5-Difluoro-4-methylphenol
Formula: C₇H₆F₂O
MW: 144.119 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18394888
CAS Number: 2985-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR