4-Bromo-2-chloro-1-(difluoromethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

35789-1G 35789-25G 35789-500MG 35789-5G
76915-510EA 290.69 USD
76915-510 76915-512 76915-514 76915-516
4-Bromo-2-chloro-1-(difluoromethoxy)benzene ≥95%
4-Bromo-2-chloro-1-(difluoromethoxy)benzene
Formula: C₇H₄BrClF₂O
Storage Temperature: Refrigerator
MDL Number: MFCD14698392
CAS Number: 1062614-42-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR