3-Bromo-5-oxazol-5-yl-pyridine ≥97%

Supplier: AOBChem USA

Warning

35707-1G 35707-25G 35707-500MG 35707-5G
76915-496EA 290.69 USD
76915-496 76915-498 76915-500 76915-502
3-Bromo-5-oxazol-5-yl-pyridine ≥97%
3-Bromo-5-oxazol-5-yl-pyridine
Formula: C₈H₅BrN₂O
MW: 225.042 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18255386
CAS Number: 1256819-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR