4-(4-Nitrophenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

35598-1G 35598-25G 35598-500MG 35598-5G
76915-472EA 165.51 USD
76915-472 76915-474 76915-476 76915-478
4-(4-Nitrophenyl)pyridine ≥97%
4-(4-Nitrophenyl)pyridine
Formula: C₁₁H₈N₂O₂
MW: 200.193 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00450018
CAS Number: 4282-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR