3-Amino-6-bromo-2-chloro-4-methylpyridine ≥97% research grade

Supplier: AOBChem USA

Warning

35555-1G 35555-25G 35555-500MG 35555-5G
76915-456EA 242.95 USD
76915-456 76915-458 76915-460 76915-462
3-Amino-6-bromo-2-chloro-4-methylpyridine ≥97% research grade
3-Amino-6-bromo-2-chloro-4-methylpyridine
Formula: C₆H₆BrClN₂
Storage Temperature: Ambient
MDL Number: MFCD11501120
CAS Number: 1038920-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR