3-Bromo-5-chloro-4-iodotoluene ≥97%

Supplier: AOBChem USA

Warning

35492-1G 35492-25G 35492-500MG 35492-5G
76915-416EA 379.81 USD
76915-416 76915-418 76915-420 76915-422
3-Bromo-5-chloro-4-iodotoluene ≥97%
3-Bromo-5-chloro-4-iodotoluene
Formula: C₇H₅BrClI
MW: 331.376 g/mol
Storage Temperature: Ambient
CAS Number: 1774897-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR