3,5-Dibromo-4-methylbenzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

35427-1G 35427-25G 35427-500MG 35427-5G
76915-402EA 260.99 USD
76915-402 76915-404 76915-406 76915-408
3,5-Dibromo-4-methylbenzaldehyde ≥97% research grade
3,5-Dibromo-4-methylbenzaldehyde
Formula: C₈H₆Br₂O
MW: 277.941 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00277892
CAS Number: 248607-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR