4-(Hydroxymethyl)-2,6-dimethylphenol ≥97%

Supplier: AOBChem USA

Warning

35248-1G 35248-25G 35248-500MG 35248-5G
76915-380EA 171.87 USD
76915-380 76915-382 76915-384 76915-386
4-(Hydroxymethyl)-2,6-dimethylphenol ≥97%
4-(Hydroxymethyl)-2,6-dimethylphenol
Formula: C₉H₁₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD02912490
CAS Number: 4397-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR