2-Hydroxy-5-(4'-fluorophenyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(4-Fluorophenyl)-2-formylphenol

35029-1G 35029-25G 35029-5G
76915-300EA 99.73 USD
76915-300 76915-302 76915-304
2-Hydroxy-5-(4'-fluorophenyl)benzaldehyde ≥97%
2-Hydroxy-5-(4'-fluorophenyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₉FO₂
MW: 216.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06802113
CAS Number: 847754-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR