3-Chloro-6-fluoro-2-hydroxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

34962-1G 34962-500MG 34962-5G
76915-286EA 544.25 USD
76915-286 76915-288 76915-290
3-Chloro-6-fluoro-2-hydroxybenzoic acid ≥97%
3-Chloro-6-fluoro-2-hydroxybenzoic acid
Formula: C₇H₄ClFO₃
MW: 190.556 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22415146
CAS Number: 1784624-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR