3-Chloro-4-cyclopropylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

34794-1G 34794-500MG 34794-5G
76915-230EA 647.15 USD
76915-230 76915-232 76915-234
3-Chloro-4-cyclopropylbenzaldehyde ≥97%
3-Chloro-4-cyclopropylbenzaldehyde
Formula: C₁₀H₉ClO
MW: 180.631 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28642190
CAS Number: 1691650-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR