3-Chloro-4-phenoxybenzoic acid methyl ester ≥97%

Supplier: AOBChem USA

Warning

34710-1G 34710-500MG 34710-5G
76915-174EA 524.09 USD
76915-174 76915-176 76915-178
3-Chloro-4-phenoxybenzoic acid methyl ester ≥97%
3-Chloro-4-phenoxybenzoic acid methyl ester
Formula: C₁₄H₁₁ClO₃
MW: 262.688 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31696100
CAS Number: 158771-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR