4-(3-Thienyl)benzenecarbaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(Thien-3-yl)benzaldehyde

34478-1G 34478-25G 34478-5G
76915-104EA 96.55 USD
76915-104 76915-106 76915-108
4-(3-Thienyl)benzenecarbaldehyde ≥97%
4-(3-Thienyl)benzenecarbaldehyde
Formula: C₁₁H₈OS
MW: 188.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039151
CAS Number: 157730-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR