3-Methyl-4-phenylphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

34413-1G 34413-25G 34413-500MG 34413-5G
76915-076EA 381.93 USD
76915-076 76915-078 76915-080 76915-082
3-Methyl-4-phenylphenylboronic acid ≥95%
3-Methyl-4-phenylphenylboronic acid
Formula: C₁₃H₁₃BO₂
Storage Temperature: Refrigerator
MDL Number: MFCD28098195
CAS Number: 1029439-30-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR