3,4-Dichloro-2-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

34330-1G 34330-25G 34330-500MG 34330-5G
76915-044EA 409.51 USD
76915-044 76915-046 76915-048 76915-050
3,4-Dichloro-2-fluorobenzoic acid ≥97%
3,4-Dichloro-2-fluorobenzoic acid
Formula: C₇H₃Cl₂FO₂
Storage Temperature: Refrigerator
MDL Number: MFCD19686190
CAS Number: 1349715-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR