3,5-Dichloro-2-pyridone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3,5-Dichloro-2-hydroxypyridine

34319-25G 34319-5G
76915-034EA 354.35 USD
76915-034 76915-036
3,5-Dichloro-2-pyridone ≥97%
3,5-Dichloro-2-pyridone

Precaución: for Research Use Only

Formula: C₅H₃Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD00015401
CAS Number: 5437-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR