3-Bromo-4-methylpyridine-5-boronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

34250-1G 34250-500MG 34250-5G
76914-982EA 587.74 USD
76914-982 76914-984 76914-986
3-Bromo-4-methylpyridine-5-boronic acid pinacol ester ≥95%
3-Bromo-4-methylpyridine-5-boronic acid pinacol ester
Formula: C₁₂H₁₇BBrNO₂
MW: 297.984 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26519033
CAS Number: 2043397-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR