4-Bromo-1-cyclopentyl-2-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

33894-1G 33894-25G 33894-500MG 33894-5G
76914-904EA 439.22 USD
76914-904 76914-906 76914-908 76914-910
4-Bromo-1-cyclopentyl-2-fluorobenzene ≥97%
4-Bromo-1-cyclopentyl-2-fluorobenzene
Formula: C₁₁H₁₂BrF
MW: 243.115 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28387957
CAS Number: 1776923-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR