3-Cyano-2,4-difluorobenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
33753-1G
33753-25G
33753-500MG
33753-5G
76914-880EA
300.24
USD
76914-880
76914-882
76914-884
76914-886
3-Cyano-2,4-difluorobenzoic acid ≥97%
3-Cyano-2,4-difluorobenzoic acid
Formula:
C₈H₃F₂NO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD20645230 CAS Number: 177942-39-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...