3-Cyano-2,4-dichlorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

33700-1G 33700-25G 33700-500MG 33700-5G
76914-868EA 260.99 USD
76914-868 76914-870 76914-872 76914-874
3-Cyano-2,4-dichlorobenzaldehyde ≥97%
3-Cyano-2,4-dichlorobenzaldehyde
Formula: C₈H₃Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD31561315
CAS Number: 2145093-86-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR