3-Bromo-2,6-dichlorobenzonitrile ≥95%

Supplier: AOBChem USA

Warning

33692-1G 33692-25G 33692-500MG 33692-5G
76914-854EA 260.99 USD
76914-854 76914-856 76914-858 76914-860
3-Bromo-2,6-dichlorobenzonitrile ≥95%
3-Bromo-2,6-dichlorobenzonitrile
Formula: C₇H₂BrCl₂N
Storage Temperature: Refrigerator
MDL Number: MFCD29043871
CAS Number: 1421620-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR