3-Bromo-2,6-difluorobenzamide ≥97%

Supplier: AOBChem USA

Warning

33680-1G 33680-25G 33680-500MG 33680-5G
76914-842EA 231.28 USD
76914-842 76914-844 76914-846 76914-848
3-Bromo-2,6-difluorobenzamide ≥97%
3-Bromo-2,6-difluorobenzamide

Precaución: for Research Use Only

Formula: C₇H₄BrF₂NO
Storage Temperature: Refrigerator
MDL Number: MFCD26680221
CAS Number: 1541810-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR