3-Fluoro-4-methyl-2-(methylthio)phenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

33647-1G 33647-25G 33647-500MG 33647-5G
76914-812EA 203.7 USD
76914-812 76914-814 76914-816 76914-818
3-Fluoro-4-methyl-2-(methylthio)phenylboronic acid ≥97% research grade
3-Fluoro-4-methyl-2-(methylthio)phenylboronic acid
Formula: C₈H₁₀BFO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD31382563
CAS Number: 2121512-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR