3-Bromo-5-butoxyphenylboronic acid, pinacol ester ≥97%

Supplier: AOBChem USA

Warning

33604-1G 33604-25G 33604-5G
76914-798EA 94.43 USD
76914-798 76914-800 76914-802
3-Bromo-5-butoxyphenylboronic acid, pinacol ester ≥97%
3-Bromo-5-butoxyphenylboronic acid, pinacol ester
Formula: C₁₆H₂₄BBrO₃
MW: 355.075 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12546590
CAS Number: 1218790-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR