(3-Bromo-5-propoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-5-propoxyphenylboronic acid

33601-25G 33601-5G
76914-790EA 204.76 USD
76914-790 76914-792
(3-Bromo-5-propoxyphenyl)boronic acid ≥97%
(3-Bromo-5-propoxyphenyl)boronic acid
Formula: C₉H₁₂BBrO₃
MW: 258.90 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07784394
CAS Number: 871126-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR