3-Chloro-5-fluoro-2-methoxybenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

33535-1G 33535-25G 33535-500MG 33535-5G
76914-742EA 195.21 USD
76914-742 76914-744 76914-746 76914-748
3-Chloro-5-fluoro-2-methoxybenzyl alcohol ≥97%
3-Chloro-5-fluoro-2-methoxybenzyl alcohol
Formula: C₈H₈ClFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD28523079
CAS Number: 1785513-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR