3-Bromo-2-fluoro-5-(trifluoromethyl)phenol ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
33501-1G
33501-25G
33501-500MG
33501-5G
76914-712EA
155.96
USD
76914-712
76914-714
76914-716
76914-718
3-Bromo-2-fluoro-5-(trifluoromethyl)phenol ≥97% research grade
3-Bromo-2-fluoro-5-(trifluoromethyl)phenol
Formula:
C₇H₃BrF₄O MW: 258.996 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD28739064 CAS Number: 1807005-89-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...