3-Bromo-2-fluoro-5-trifluoromethylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

33499-25G 33499-5G
76914-700EA 595.17 USD
76914-700 76914-702
3-Bromo-2-fluoro-5-trifluoromethylphenylboronic acid ≥97%
3-Bromo-2-fluoro-5-trifluoromethylphenylboronic acid
Formula: C₇H₄BBrF₄O₂
MW: 286.814 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20441821
CAS Number: 2096338-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR