3-Bromo-2-fluoro-5-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

33496-1G 33496-25G 33496-500MG 33496-5G
76914-684EA 134.74 USD
76914-684 76914-686 76914-688 76914-690
3-Bromo-2-fluoro-5-(trifluoromethyl)benzoic acid ≥97%
3-Bromo-2-fluoro-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₃BrF₄O₂
MW: 287.006 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31382551
CAS Number: 2091606-48-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR