3-Chloro-2,4-difluorobenzonitrile ≥97% research grade

Supplier: AOBChem USA

Warning

33433-1G 33433-25G 33433-5G
76914-676EA 76.39 USD
76914-676 76914-678 76914-680
3-Chloro-2,4-difluorobenzonitrile ≥97% research grade
3-Chloro-2,4-difluorobenzonitrile
Formula: C7H2ClF2N
MW: 173.5500 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06658267
CAS Number: 887267-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR