4-(Benzyloxy)-2,6-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

33262-1G 33262-25G 33262-500MG 33262-5G
76914-662EA 210.06 USD
76914-662 76914-664 76914-666 76914-668
4-(Benzyloxy)-2,6-dimethylbenzaldehyde ≥97%
4-(Benzyloxy)-2,6-dimethylbenzaldehyde
Formula: C₁₆H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD06797237
CAS Number: 28924-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR